Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Plakophilin 1 Mouse anti-Human, Mouse, Clone: 10B2, Novus Biologicals™

Mouse Monoclonal Antibody

$152.22 - $378.35

Specifications

Antigen Plakophilin 1
Clone 10B2
Host Species Mouse
Isotype IgG1
Gene Alias B6PBand 6 protein, MGC138829, plakophilin 1 (ectodermal dysplasia/skin fragility syndrome), plakophilin-1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB25332620 View Documents Novus Biologicals
NBP25332620UG
20 ug Each for $152.22
Add to cart
 
NB253326100 View Documents Novus Biologicals
NBP253326100UG
100 ug Each for $378.35
Add to cart
 
NB253326200 View Documents Novus Biologicals
NBP253326200UG
200 ug This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Plakophilin 1 Monoclonal antibody specifically detects Plakophilin 1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, SDS-Page.
Specifications

Specifications

Plakophilin 1
Mouse
B6PBand 6 protein, MGC138829, plakophilin 1 (ectodermal dysplasia/skin fragility syndrome), plakophilin-1
Flow Cytometry, Immunocytochemistry, Immunofluorescence, SDS-Page, Western Blot
Unconjugated
Store at 4C.
Monoclonal
0.2 mg/mL
10B2
IgG1
Human, Mouse
RUO
Purified
Primary
10mM PBS with 0.05% BSA with 0.05% Sodium Azide

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit