Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Plant Life Cycle Lab Investigation
Fascinating, hands-on exploration of plant germination and growth.
Specifications
Class Size | 40 students |
---|---|
For Use With (Application) | Study the fundamentals of plant growth and anatomy |
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
S32665
![]() |
Neo/Sci Corporation
201363 |
Plant Life Cycle Lab Investigation Kit |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
- Experience the fundamentals of plant growth and anatomy
- Germinate seeds
- Study the stages of plant growth, processes of seed germination, seedling development and anatomy of the plant embryo, root and stem
- Curriculum pack features kit plus a corresponding CD-ROM (S66711)
Includes further investigations for exploring:
- Effects of light
- Temperature
- pH
- Environmental pollutants
Specifications
40 students | |
Study the fundamentals of plant growth and anatomy |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title