Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Plant Rubisco ELISA Kit (Colorimetric)

Assay has high sensitivity and specificity for detection of plant RuBisCO

$732.50

Specifications

Sample Volume 50 to 100 μL
Kit Contents Species, Sensitivity, SampleType, ResearchDiscipline, Specificity, Synonym, StorageRequirements, Target, ForUseWithApplication, dimProductType, AssayRange, SampleVolume, KitContents
Sensitivity 1.56μg/mL
Sample Type Plant Tissue
Research Discipline Cell Biology
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB601421KT View Documents Novus Biologicals™
NBP2601421KIT
1 Kit Each for $732.50
Add to cart
 
Description

Description

The Plant Rubisco ELISA Kit (Colorimetric) measures plant Rubisco in plant tissues.
Specifications

Specifications

50 to 100 μL
1.56μg/mL
Cell Biology
Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
ELISA
3.12μg/mL to 800μg/mL
Species, Sensitivity, SampleType, ResearchDiscipline, Specificity, Synonym, StorageRequirements, Target, ForUseWithApplication, dimProductType, AssayRange, SampleVolume, KitContents
Plant Tissue
For detection of plant RuBisCO
Rubisco
ELISA Kit (Colorimetric)

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit