Learn More
BioVision IncSupplier Diversity Partner Plant Tissue Extraction Kit: Simple method for extracting soluble protein, biomolecules & metabolites from plant tissues without the use of liquid nitrogen. Protein yield ~ 2-9 mg/ml depending on the sample type. 50 preps. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K29650
Biovision’s Plant Protein Extraction Kit provides an effective way of extracting soluble protein and other biomolecules and metabolites from all kinds of dry and fresh plant tissues without the use of liquid nitrogen. The kit includes optimized Extraction Buffer to extract plant protein and metabolites, Protease Inhibitor Cocktail to prevent protein degradation and Homogenization Pestles and Tubes to homogenize the plant tissue. The extracted protein can be used in a variety of downstream applications such as Western Blotting, 2-D gel electrophoresis, immunoprecipitation assay, and enzymatic activity assays etc. and extracted small molecules can be used in plant metabolic studies. This easy-to-use and rapid procedure ensures consistent protein yield ~ 2-9 mg/ml depending on the sample type. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9672