Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Plant Vitamin B12 ELISA Kit (Colorimetric)

Assay has high sensitivity and specificity for detection of plant Vitamin B12

$732.50

Specifications

Sensitivity 14.967pg/mL
Sample Type Plant Tissue
Research Discipline Lipid and Metabolism
Specificity For detection of plant VB12
Storage Requirements Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB601971KT View Documents Novus Biologicals™
NBP2601971KIT
1 Kit Each for $732.50
Add to cart
 
Description

Description

The Plant Vitamin B12 ELISA Kit (Colorimetric) measures plant Vitamin B12 in plant tissue.
Specifications

Specifications

14.967pg/mL
Lipid and Metabolism
Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
ELISA
15.6 pg/ml to 1000 pg/ml
Instruction manual, Standard, Sample Diluent, Stop Solution, TMB Substrate, Wash Buffer (25 x concentrate), HRP-avidin Diluent, HRP-avidin (100 x concentrate), Assay plate, Biotin-conjugate (10 x concentrate), Biotin-conjugate Diluent, Adhesive Strip (For 96 wells)
Plant Tissue
For detection of plant VB12
Vitamin B12
ELISA Kit (Colorimetric)
50 to 100 μL

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit