Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Plas Labs™ Animal Operating Boards Available on GSA/VA Contract for Federal Government customers only.
Useful for surgery, biopsy, necropsy, injections and basic restraint
$361.50 - $399.00
Specifications
Color White
Type Animal Operating Board
Includes 5 adjustable nylon cords
Material Noryl™ Plastic
View More Specs
Includes: Five adjustable nylon cords
Products 2
Catalog Number Mfr. No. Length (English) Length (Metric) Width (English) Width (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Length (Metric) Width (English) Width (Metric) Price Quantity & Availability  
01-288-30A Available on GSA/VA Contract for Federal Government customers only.
View Documents
Plas Labs™
508MS
11 in. 28 cm 4.5 in. 11.43 cm
Each for $361.50
N/A
01-330
View Documents
Plas Labs™
510SS
16 in. 40.64 cm 8 in. 20.32 cm
Each for $399.00
N/A
Description
Description
  • One-piece Noryl™ plastic boards accommodate small animals
  • Suction cup feet secure 01-288-30A
  • Metal bracket on 01-330 enables user to elevate the head end of board if needed
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
White
Animal Operating Board
5 adjustable nylon cords
Noryl™ Plastic