Plas Labs™ Model 830 Compact Glove Box

For safe containment of hazardous materials when being weighed

Manufacturer: Plas Labs™ 830ABE

 View all options for this product

Catalog No. 11-386-137

  Check Availability
Add to Cart


    Voltage 120V
    Certifications/Compliance UL, CSA, CE
    Depth (English) Interior 29 in.
    Depth (Metric) Exterior 81.2cm
    Depth (Metric) Interior 74cm
    Depth (English) Exterior 32 in.
    Type Glove Box
    Height (English) Exterior 35 in.
    Width (English) Exterior 76 in.
    Height (English) Interior 32 in.
    Height (Metric) Exterior 89cm
    Width (English) Interior 60 in.
    Width (Metric) Exterior 193cm
    Width (Metric) Interior 152.4cm
    Dimensions (L x W x H) Exterior 32 x 76 x 35 in. (81.2 x 193 x 89cm)
    Length (English) Exterior 32 in.
    Length (Metric) Exterior 81.2cm

    Work chamber accommodates analytical and toploading balances, bench scales and moisture analyzers in a clean, safe, portable and draft-free atmosphere.

    • Top section is removable for easy installation of equipment
    • Four purging gas valves (two on the transfer chamber and two on the main chamber)
    • White leveling tray for easy transferring of liquids
    • All clamps stainless steel and completely adjustable
    • Pressure relief valve minimizes glove fight back
    • HEPA filter on relief valve prevents escape of particulates and organic vapors
    • HEPA and organic vapor canister filters (replaceable)
    • Roomy transfer chamber is 12L x 11 in. I.D.

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit