Learn More
Epredia™ Lab Vision™ Plasma Cell Ab-1 Mouse Monoclonal Antibody, 50μg/mL, BSA and azide
Provide accurate, reproducible results with the Epredia™ Plasma Cell Ab-1, Mouse Monoclonal Antibody.
Supplier: Epredia™ MS488R1
Description
Host Species: Mouse
Clone: LIV3G11
Isotype: IgG2a
Species Reactivity: Human. Does not react with rat. Others not known.
Epitope: Not determined
Positive Control: Tonsil or lymph node
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin only)
Note:
USA: RUO; Int'l: RUO
Specifications
Plasma Cell Ab-1 | |
Monoclonal | |
Unconjugated | |
Mouse | |
RUO | |
Primary | |
Prediluted |
Immunohistochemistry (Paraffin) | |
LIV3G11 | |
Ready-to-use for IHC (UVLP) | |
1 mL | |
Hematopoietic | |
Human | |
IgG2a |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.