Learn More
Plasminogen Mouse anti-Human, DyLight 488, Clone: 5H3, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB1107926G
Catalog No. NB1107926G
Add to cart
Description
Description
Plasminogen Monoclonal specifically detects Plasminogen in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Sandwich ELISA.
Specifications
Specifications
Plasminogen
5H3
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Sandwich ELISA
Purified
PLG
Full length native protein (purified) (Human).
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
Reacts with plasminogen, plasmin and angiostatin.
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
DKFZp779M0222, EC 3.4.21, EC 3.4.21.7, plasmin, plasminogen
Mouse
IgG1
0.2 ml
Angiogenesis, Cancer, Cell Cycle and Replication
Primary
5340
Human