Learn More
Mopec Plastic Grey X-Ray Autopsy Cart
Constructed of vacuum formed ABS plastic, making it an incredibly durable and lightweight body tray that is ideal for any medical examiners office. Mopec Plastic Grey X-Ray Autopsy Cart Top is designed to universally interface with any line of autopsy carts and body racking systems.
Manufacturer: Mopec GA850
Description
- Can be used with cantilever rack storage systems as well as conveyor rack system
- Durable and lightweight
- Gray in color
- Rails to hold X-Ray cassettes
- Recessed channels for improved drainage of fluids
- Includes a drain stopper and safety chain
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Warranty and Services
1 Year Warranty
Specifications
Autopsy Cart | |
Plastic | |
32.1 in. | |
209.55cm | |
12.7cm |
Plastic Grey X-Ray | |
82.5 in. | |
5 in. | |
81.53cm | |
Drain Stopper and Safety Chain |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.