Learn More
Sklar InstrumentsSupplier Diversity Partner Plastic Procedure Tray, Sklar, 10 1/8 in. x 6 5/8 in., Plastic. Sklar's Plastic Procedure Trays are used to sterilize, store, transport, and arrange medical instruments. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 101960
Plastic Procedure Tray, Sklar, 10 1/8 in. x 6 5/8 in., Plastic. Sklar's Plastic Procedure Trays are used to sterilize, store, transport, and arrange medical instruments. The trays are available in perforated and non-perforated styles and multiple sizes. They feature a texturized surface to help prevent instruments from slipping.
Catalog No. 50-143-7157