Learn More
Action Pump Plastic Siphon Drum Pumps
Choice of polyethylene or polypropylene models
Supplier:  Action Pump 4009
 View more versions of this product
Catalog No. 01-007-2
Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative.
Description
Description
Choice of polyethylene or polypropylene models
  • For dispensing of dilute acids, caustics, oils, waxes, disinfectants without leakage problems.
  • Maximum delivery rate varies with model
  • Use for 15-30 and 55 gal. (57-114, 208L) drums with a 2 in. (5.1cm) bung
  • Some units can be used with 5 gal. pail with separate adapter
  • Heavy-duty model available
Specifications
Specifications
Polyethylene
40°F
Dispensing of dilute acids, caustics, oils, waxes, disinfectants without leakage problems
Buttress Siphon Pump
140°F
15 to 30 or 55 gal. drum and 5 gal. pail with separate adapter