Thermo Scientific™ Plastic Spill-Free Slide Jar

Hold 10 slides, one per slot with the Thermo Scientific™ Plastic Spill-Free Slide Jar.

Manufacturer: Thermo Scientific™ 1001363

 View more versions of this product

Catalog No. 10-013-63

  Check Availability
Add to Cart

    Specifications

    Width (English) 1 in.
    Width (Metric) 26mm
    Item Description 12/Pack
    Quantity 12/Pack
    Length (English) 1.4 in.
    Length (Metric) 36mm
    Slide Count 10

    Hold 10 slides, one per slot with the Thermo Scientific™ Plastic Spill-Free Slide Jar.

    • 1.4 x 1 in. (36 x 26mm) rectangular top with straight body
    • Polypropylene construction with internal grooves
    • Integral snap-lock lid with reagent-proof lid seal
    • Microwavable

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit