Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Plastic Warning Sign: FLAMMABLE MATERIAL. KEEP FIRE AWAY.

Improve workplace safety with proper signage and markings. This Brady™ corner hole mounted sign is a durable, clear and effective tool to raise awareness to chemical, biohazard and hazardous material warnings.

Manufacturer:  Brady™ 26553

Catalog No. 19-106-945


Add to cart

Description

Description

  • Bold and bright, providing high visibility to critical messages
  • For indoor and outdoor use
Specifications

Specifications

Black/red on white
PS (Polystyrene)
Rectangle
Material: Laminated polystyrene
10 in.
FLAMMABLE MATERIAL. KEEP FIRE AWAY.
0.0591 in.
14 in.
Not Laminated
1/Ea.
14 in. x 10 in. x 0.0591 in.
Corner holes
176°F
-40°F
Non-adhesive
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit