Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Plastin L Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP294267100UL
Description
Plastin L Polyclonal antibody specifically detects Plastin L in Human, Mouse samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
Plastin L | |
Polyclonal | |
PBS (pH 7.3), 50% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
3936 | |
Store at -20°:C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunoprecipitation | |
Unconjugated | |
CP64, DKFZp781A23186, FLJ25423, FLJ26114, FLJ39956, LC64PPLS2bA139H14.1 (lymphocyte cytosolic protein 1 (L-plastin)), LCP-1, LPL, L-plastin, L-plastin (Lymphocyte cytosolic protein 1) (LCP-1) (LC64P), Lymphocyte cytosolic protein 1, lymphocyte cytosolic protein 1 (L-plastin), Lymphocyte cytosolic protein-1 (plasmin), plastin 2, plastin-2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human LCP1 (NP_002289.2). | |
100 μg | |
Primary | |
Human, Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title