Learn More
BiochromatoSupplier Diversity Partner PLATE SEAL SLIT SEAL 1 BOX DFS Item
Manufacturer:  BiochromatoSupplier Diversity Partner R80.120.00
Slit Seal from BioChromato are resistant, instant self-closing 96-well plate seals. Used for prevention of solvent evaporation and cross-contamination.
Catalog No. 50-103-7298