Learn More
Corning™ Platelet-Derived Growth Factor-BB (PDGF-BB)
Designed specifically for use in basic cell biology research
Manufacturer:  Corning™ 356051
Catalog No. MT356051
Add to cart
Description
Description
  • A dimeric glycoprotein composed of two β chains and is a mitogen in serum for mesenchymal cells
  • The human recombinant form is isolated from E. coli
  • Products are tested for biological activity in appropriate assays, such as the ability of growth factors to stimulate proliferation or DNA synthesis
  • Lymphokines and growth factors are also sterile filtered generally using a 0.2µm membrane
  • Tested for the presence of bacteria, fungi and mycoplasma
Specification sheets include detailed information on quality control testing, product source, storage, stability and recommendations on proper experimental concentrations.
Specifications
Specifications
10 x 10 μg
2-8°C
Human