Learn More
Thermo Scientific™ Plates and Modules with Affinity Binding Surfaces Available on GSA/VA Contract for Federal Government customers only.
Help reduce variability in molecular orientation with Thermo Scientific™ Plates and Modules with Affinity Binding Surfaces.
Manufacturer:  Thermo Scientific™ 436032
 View more versions of this product
Catalog No. 12-566-91
  Check Availability
Add to cart
Disclaimers:
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description
Help reduce variability in molecular orientation with Thermo Scientific™ Plates and Modules with Affinity Binding Surfaces.
Affinity Capture Surfaces (Passive Streptavidin and Immobilizer Streptavidin, Nickel Chelate and Glutathione, BioBind)
  • Highly specific binding
  • Reduced variability in molecular orientation
  • Immobilizer surfaces improve signal to noise ratios
  • Streptavidin biotin interaction can be exploited to immobilize a wide range of biomolecules
Description & Specifications
Specifications
Affinity
96-well plate
1
Solid plate 96-well
100µL
Unspecified
Clear
15
Glutathione
400µL
Flat
Colorimetric