Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Platinum, 0.1% on 2-4mm alumina spheres

VOC removal from vent streams by combination with O2

Manufacturer:  Alfa Aesar™ 4220522

Quantity 100g

 View more versions of this product

Catalog No. AA4220522


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

MFCD00011179
Platinum, 0.1% on 2 to 4mm alumina spheres

Specifications

Spheres
MFCD00011179
100g
Platinum, 0.1% on 2 to 4mm alumina spheres

Safety and Handling

SDS & Safety Documents

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit