Alfa Aesar™ Platinum, 0.5% on 2.7-3.3mm (0.11-0.13in) alumina pellets, reduced

Manufacturer: Alfa Aesar™ 8910622

Quantity 100g

 View more versions of this product

Catalog No. AA8910622

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    MDL Number MFCD00011179
    Chemical Name or Material Platinum, 0.5% on alumina pellets, 2.7 to 3.3mm (0.11 to 0.13 in.), Reduced

    Specifications

    TSCA TSCA
    MDL Number MFCD00011179
    Quantity 100g
    Chemical Name or Material Platinum, 0.5% on alumina pellets, 2.7 to 3.3mm (0.11 to 0.13 in.), Reduced

    Description

    • Selective hydrogenations; gas purifications: e
    g
  • Oxidation of carbon monoxide to carbon dioxide, hydrogen removal from oxygen or carbon dioxide

SDS & Safety Documents

SDS
  • Description & Specifications
  • Safety and Handling
  • Related Products


    Product Content Feedback

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit