Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Platinum, 1% on granular carbon, reduced, nominally 50% water wet

Click to view available options
Quantity:
25g
100g
7440-6-6
MFCD00011179
7440-6-6
MFCD00011179
Quantity 100g
Mesh Size 4x10 Mesh
Color Gray
Solubility Information Soluble in aqua regia.. Insoluble in water and acid.
Physical Form 4x10 Mesh
Chemical Name or Material Platinum, 1% on activated carbon powder, Reduced, nominally 50% water wet

P210-P240-P241-P280-P370+P378q

H228

TSCA : Yes

Recommended Storage : Ambient temperatures