Alfa Aesar™ Platinum, 1% on polyethyleneimine/SiO2 powder

Reduction reactions

Manufacturer: Alfa Aesar™ 4340806

Quantity 5g

 View more versions of this product

Catalog No. AA4340806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-06-4
    MDL Number MFCD00011179
    Chemical Name or Material Platinum, 1% on polyethyleneimine/SiO2

    Specifications

    Health Hazard 3 GHS P Statement
    P261-P304+P340-P312-P405-P403+P233-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Store locked up.
    Store in a well-ventilated place.
    Keep container tightly closed.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Physical Form Powder
    Health Hazard 1 Warning
    Mesh Size 40 to 200 Mesh
    MDL Number MFCD00011179
    CAS 7440-06-4
    Odor Odorless
    Health Hazard 2 GHS H Statement
    H332-H335
    Harmful if inhaled.
    May cause respiratory irritation.
    Quantity 5g
    Chemical Name or Material Platinum, 1% on polyethyleneimine/SiO2

    GHS H Statement
    H332-H335
    Harmful if inhaled.
    May cause respiratory irritation.

    GHS P Statement
    P261-P304+P340-P312-P405-P403+P233-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Store locked up.
    Store in a well-ventilated place.
    Keep container tightly closed.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit