Alfa Aesar™ Platinum, 10% on alumina, reduced

Manufacturer: Alfa Aesar™ 4449414

Quantity 25g

 View more versions of this product

Catalog No. AA4449414

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00011179
    Chemical Name or Material Platinum, 10% on alumina, Reduced

    Specifications

    Health Hazard 3 GHS P Statement
    P261-P281-P304+P340-P312-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 1 Warning
    MDL Number MFCD00011179
    Health Hazard 2 GHS H Statement
    H341-H335
    Suspected of causing genetic defects.
    May cause respiratory irritation.
    Quantity 25g
    Physical Form Granular
    Chemical Name or Material Platinum, 10% on alumina, Reduced

    GHS H Statement
    H341-H335
    Suspected of causing genetic defects.
    May cause respiratory irritation.

    GHS P Statement
    P261-P281-P304+P340-P312-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit