Alfa Aesar™ Platinum 13 wt% Rhodium wire, 0.203mm (0.008 in.) dia., ISA Type R Standard Grade Thermocouple

Manufacturer: Alfa Aesar™ 42905G1

Quantity 1m

 View more versions of this product

Catalog No. AA42905G1

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00198186
    Chemical Name or Material Platinum 13 wt% Rhodium wire, 0.203mm (0.008 in.) dia., ISA Type R Standard Grade Thermocouple

    Specifications

    MDL Number MFCD00198186
    Odor Odorless
    Weight ≈0.64g/m
    TSCA TSCA
    Avg. Mol. Wt. or Mol. Wt. Range Pt:Rh; 87:13 wt%
    Quantity 1m
    Physical Form Wire
    Chemical Name or Material Platinum 13 wt% Rhodium wire, 0.203mm (0.008 in.) dia., ISA Type R Standard Grade Thermocouple

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit