Alfa Aesar™ Platinum, 3% on activated carbon powder, standard, unreduced, nominally 50% water wet

Manufacturer: Alfa Aesar™ 3831706

Quantity 5g

 View more versions of this product

Catalog No. AA3831706

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00011179
    Chemical Name or Material Platinum, 3% on activated carbon powder, Standard, unreduced, nominally 50% water wet

    Specifications

    MDL Number MFCD00011179
    Weight 900 to 1100m2/g
    TSCA TSCA
    Quantity 5g
    Chemical Name or Material Platinum, 3% on activated carbon powder, Standard, unreduced, nominally 50% water wet

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit