Alfa Aesar™ Platinum 30 wt% Rhodium wire, 0.33mm (0.013 in.) dia., ISA Type B Standard Grade Thermocouple

Manufacturer: Alfa Aesar™ 45571G2

Quantity 2m

 View more versions of this product

Catalog No. AA45571G2

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00198186
    Chemical Name or Material Platinum 30 wt% Rhodium wire, 0.33mm (0.013 in.) dia., ISA Type B Standard Grade Thermocouple

    Specifications

    Odor Odorless
    TSCA TSCA
    Physical Form Wire
    MDL Number MFCD00198186
    Avg. Mol. Wt. or Mol. Wt. Range Pt:Rh; 70:30 wt%
    Weight ≈0.75g/50cm
    Quantity 2m
    Chemical Name or Material Platinum 30 wt% Rhodium wire, 0.33mm (0.013 in.) dia., ISA Type B Standard Grade Thermocouple

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit