Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Platinum on activated carbon, 10% Pt, ca .50% moisture, ACROS Organics™

Click to view available options
Quantity:
1g
10g
50g
Packaging:
Glass bottle
7440-06-4
Pt
195.08
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
black,platin,sponge,platinum, metal,platinum, elemental,platine,platino,metal,platin german,iv ion
23939
CHEBI:33400
platinum
[Pt]
7440-06-4
Pt
195.08
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
black,platin,sponge,platinum, metal,platinum, elemental,platine,platino,metal,platin german,iv ion
23939
CHEBI:33400
platinum
[Pt]
Quantity 50g
Loss on Drying 45% to 55% (typically)
Packaging Glass bottle
Formula Weight 195.08
Physical Form Powder
Percent Purity 10% (Pt)
Color Black
Chemical Name or Material Platinum on activated carbon

GHS H Statement:
Self-heating in large quantities; may catch fire.

GHS P Statement:
Keep cool. Protect from sunlight.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning