Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Platinum on activated carbon, 5% Pt, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
7440-06-4
Pt
195.08
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
black,platin,sponge,platinum, metal,platinum, elemental,platine,platino,metal,platin german,iv ion
23939
CHEBI:33400
platinum
[Pt]
7440-06-4
Pt
195.08
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
black,platin,sponge,platinum, metal,platinum, elemental,platine,platino,metal,platin german,iv ion
23939
CHEBI:33400
platinum
[Pt]
Quantity 50g
Fieser 01,982; 08,405; 04,393; 04,393; 04,393; 04,393
Packaging Glass bottle
Formula Weight 195.08
Physical Form Powder
Percent Purity 5% (Pt)
Particle Size Particle size distribution: <7 Micron: 10%, <28 Micron: 50%, <85 Micron: 90%
Surface Area 900m2/g
Color Black
Chemical Name or Material Platinum on activated carbon, 5% Pt