Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Platinum black, 98% Pt, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
7440-06-4
Pt
195.08
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
black,platin,sponge,platinum, metal,platinum, elemental,platine,platino,metal,platin german,iv ion
23939
CHEBI:33400
platinum
[Pt]
7440-06-4
Pt
195.08
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
black,platin,sponge,platinum, metal,platinum, elemental,platine,platino,metal,platin german,iv ion
23939
CHEBI:33400
platinum
[Pt]
Additional Information alkali (K, Na): < 0.4%
Quantity 5g
Calcium (Ca) 50ppm max.
Copper (Cu) 50ppm max.
Density 21.45
Gold (Au) 50ppm max.
Linear Formula Pt
Iridium (Ir) 50ppm max.
Iron (Fe) 50ppm max.
Fieser 01,890; 02,332
Merck Index 15, 7644
Magnesium (Mg) 50ppm max.
Packaging Glass bottle
Palladium (Pd) 50ppm max.
Formula Weight 195.08
Physical Form Powder
Percent Purity ≥97.5% (Pt)
Rhodium (Rh) 50ppm max.
Ruthenium (Ru) 50ppm max.
Silver (Ag) 50ppm max.
Specific Surface Area 25 m2/g min.
Color White
Melting Point 1772.0°C
Boiling Point 3827.0°C
Chemical Name or Material Platinum black, 98% Pt
Show More Show Less

GHS H Statement:
Flammable solid.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 231-116-1

RTECSNumber : TP2160000

TSCA : TSCA