Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Platinum black, high surface area, Alfa Aesar™

Form: S.A. 25.0-30.4m2/g

Manufacturer:  Alfa Aesar 4383803

Quantity 1g

 View more versions of this product

Catalog No. AA4383803


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7440-06-4
Powder
MFCD00011179
Platinum

Specifications

1g
Dark gray powder
Powder
Odorless
Platinum
7440-06-4
21.45g/cm3 at 20°C
MFCD00011179
Dark gray

Safety and Handling

SDS & Safety Documents

H228: Flammable solid.

DOTInformation : DOT Class: 4.1, Packaging Group: II

EINECSNumber : 231-116-1

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit