Alfa Aesar™ Platinum flake, -200 mesh, 99.7% (metals basis)

Manufacturer: Alfa Aesar™ 4341806

Quantity 5g

 View more versions of this product

Catalog No. AA4341806

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 7440-06-4
    MDL Number MFCD00011179
    Chemical Name or Material Platinum flake

    Specifications

    Quantity 5g
    Mesh Size −200 Mesh
    Odor Odorless
    TSCA TSCA
    Physical Form Flakes
    MDL Number MFCD00011179
    RTECS Number TP2160000
    CAS 7440-06-4
    Chemical Name or Material Platinum flake
    Assay 99.7%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit