Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Platinum gauze, 52 mesh woven from 0.1mm dia. wire, 99.9% (metals basis), Alfa Aesar™

Click to view available options
Quantity:
25 x 25mm
50 x 50mm
75 x 75mm
100 x 100mm
7440-06-4
Pt
195.08
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
black,platin,sponge,platinum, metal,platinum, elemental,platine,platino,metal,platin german,iv ion
23939
CHEBI:33400
platinum
[Pt]
7440-06-4
Pt
195.08
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
black,platin,sponge,platinum, metal,platinum, elemental,platine,platino,metal,platin german,iv ion
23939
CHEBI:33400
platinum
[Pt]
Quantity 100 x 100mm
Appearance Silvery white guage
Assay Percent Range 99.9%
Mesh Size 52 Mesh
Solubility Information Insoluble in water. Dissolves in aqua regia
Odor Odorless
Physical Form ≈0.47g/25x25mm,Wire Cloth
Chemical Name or Material Platinum

EINECSNumber : 231-116-1

TSCA : Yes

Recommended Storage : Ambient temperatures