Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Platinum gauze, 52 mesh woven from 0.1mm dia. wire, 99.9% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 10283M

Quantity 50 x 75mm

 View more versions of this product

Catalog No. AA10283M

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

7440-06-4
MFCD00011179
Platinum

Specifications

50 x 75mm
Silvery white guage
Gauze
MFCD00011179
Platinum
7440-06-4
21.45g/cm3 at 20°C
99.9%
Odorless

Safety and Handling

EINECSNumber : 231-116-1

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit