Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Platinum(II) acetylacetonate, 98%, ACROS Organics™
Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
Specifications
Quantity | 25g |
Color | Undesignated |
Melting Point | 250.0°C to 252.0°C |
Packaging | Glass bottle |
Sublimation Point | 170 °C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (ex Pt) |
Linear Formula | [CH3COCH=C(CH3)O]2Pt |
Solubility Information | Solubility in water: insoluble. Other solubilities: insoluble in ethanol, diethyl ether;, soluble in acetone, toluene, diluted acids |
Formula Weight | 393.31 |
Show More |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of damaging fertility or the unborn child.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Cal
GHS Signal Word: Warning
EINECSNumber : 239-223-5