Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Platinum(II) cyanide, 98+%

Click to view available options
Quantity:
0.5g
2.5g
592-06-3
C2N2Pt
MFCD00016288
platinum ii cyanide,acmc-1axcr,platinumdicarbonitrile,platinum cyanide pt cn 2 7ci,8ci,9ci,platinum ii cyanide trace metals basis
592-06-3
C2N2Pt
MFCD00016288
platinum ii cyanide,acmc-1axcr,platinumdicarbonitrile,platinum cyanide pt cn 2 7ci,8ci,9ci,platinum ii cyanide trace metals basis
Linear Formula Pt(CN)2
UN Number UN1588
Formula Weight 247.13
Percent Purity ≥98%
Quantity 2.5g
Chemical Name or Material Platinum(II) cyanide

GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CYANIDES, INORGANIC, SOLID, N.O.S.