Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Platinum Iridium wire, 0.152mm (0.006in) dia, hard, 99.9% (metals basis excluding precious metals), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 043331BW
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Platinum Iridium wire | |
Pt:Ir; 80:20 wt% | |
MFCD02091683 | |
99.9% | |
≈0.04g/10cm |
0.152mm (0.006 in.) dia., Hard | |
(metals basis excluding precious metals) | |
50 cm | |
Odorless | |
Wire |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
RUO – Research Use Only
Spot an opportunity for improvement?
Provide Content Correction