Alfa Aesar™ Platinum Iridium wire, 1.5mm (0.06 in.) dia.

Manufacturer: Alfa Aesar™ 44812BU

Quantity 25cm

 View more versions of this product

Catalog No. AA44812BU

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD02091683
    Chemical Name or Material Platinum Iridium wire, 1.5mm (0.06 in.) dia.

    Specifications

    Physical Form Wire
    Odor Odorless
    Avg. Mol. Wt. or Mol. Wt. Range Pt:Ir; 90:10 wt%
    Weight ≈1.91g/5cm
    Quantity 25cm
    TSCA TSCA
    MDL Number MFCD02091683
    Chemical Name or Material Platinum Iridium wire, 1.5mm (0.06 in.) dia.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit