Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Platinum(IV) chloride, 99.9% (metals basis), Pt 57% min, Alfa Aesar™

Manufacturer:  Alfa Aesar 1104506

Quantity 5g

 View more versions of this product

Catalog No. AA1104506


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

13454-96-1
99.9% (metals basis)
336.9
Platinum(IV) chloride, Pt 57% min.
Crystalline
PtCl4
MFCD00011182

Specifications

13454-96-1
99.9% (metals basis)
336.9
4.303g/cm3 at 20°C
5g
Crystalline
PtCl4
MFCD00011182
Red-brown
Platinum(IV) chloride, Pt 57% min.

Safety and Handling

SDS & Safety Documents

H301-H317-H334-H341-H314-H318: Toxic if swallowed. May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Suspected of causing genetic defects. Causes severe skin burns and eye damage. Causes serious eye damage.

DOTInformation : DOT Class: 8, Packaging Group: III

EINECSNumber : 236-645-1

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit