Alfa Aesar™ Platinum(IV) chloride, Premion™, 99.99+% (metals basis), Pt 57% min

Manufacturer: Alfa Aesar™ 4450303

Quantity 1g

 View more versions of this product

Catalog No. AA4450303

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 13454-96-1
    Molecular Formula PtCl4
    Formula Weight 336.9
    MDL Number MFCD00011182
    UN Number UN2923
    Chemical Name or Material Platinum (IV) chloride
    Grade Premion™

    Specifications

    Formula Weight 336.9
    Health Hazard 2 GHS H Statement
    H301-H334-H314-H318-H317
    Toxic if swallowed.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Crystalline Soluble
    Molecular Formula PtCl4
    MDL Number MFCD00011182
    CAS 13454-96-1
    Density 4.303g/mL
    Melting Point 370°C (dec.)
    Quantity 1g
    Concentration Pt 57% min
    Odor Odorless
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
    RTECS Number TP2275500
    TSCA TSCA
    Sensitivity Hygroscopic
    UN Number UN2923
    Color Brown
    Health Hazard 1 Danger
    Chemical Name or Material Platinum (IV) chloride
    Assay ≥99.99%, (metals basis)
    Grade Premion™

    GHS H Statement
    H301-H334-H314-H318-H317
    Toxic if swallowed.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause an allergic skin reaction.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit