Platinum(IV) iodide, 99.95% (Metals basis), Pt 27.3% min., Alfa Aesar™, Premion™

Manufacturer: Alfa Aesar 4040101

Quantity 0.25g

 View more versions of this product

Catalog No. AA4040101

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7790-46-7
    Physical Form Powder
    Assay Percent Range 99.95% (metals basis)
    Molecular Formula I4Pt
    Formula Weight 702.7
    MDL Number MFCD00016292
    Chemical Name or Material Platinum(IV) iodide

    Specifications

    Physical Form Powder
    Formula Weight 702.7
    Quantity 0.25g
    Color Black
    DOT Information DOT Class: 8, Packaging Group: III
    Health Hazard 2 When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
    EINECS Number 232-207-9
    Assay Percent Range 99.95% (metals basis)
    Molecular Formula I4Pt
    Appearance Black powder
    MDL Number MFCD00016292
    CAS 7790-46-7
    Health Hazard 1 H314-H318: Causes severe skin burns and eye damage. Causes serious eye damage.
    Composition Pt 27.3% min.
    Chemical Name or Material Platinum(IV) iodide

    When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

    H314-H318: Causes severe skin burns and eye damage. Causes serious eye damage.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit