Alfa Aesar™ Platinum(IV) oxide, anhydrous, Premion™, 99.95% (metals basis), Pt 84.4% min

Manufacturer: Alfa Aesar™ 4040206

Quantity 5g

 View more versions of this product

Catalog No. AA4040206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1314-15-4
    Linear Formula PtO2
    Formula Weight 227.09
    MDL Number MFCD00011184
    UN Number UN1479
    Merck Index 14,7527
    Chemical Name or Material Platinum(IV) oxide, Anhydrous
    Grade Premion™

    Specifications

    Formula Weight 227.09
    Health Hazard 3 GHS P Statement
    P221-P210-P220-P280-P370+P378a-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Keep/Store away from clothing/combustible materials.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 1314-15-4
    Health Hazard 2 GHS H Statement
    H272
    May intensify fire
    oxidizer.
    RTECS Number TP2506020
    Odor Odorless
    Melting Point 450°C
    Linear Formula PtO2
    Health Hazard 1 Danger
    TSCA TSCA
    UN Number UN1479
    Merck Index 14,7527
    Density 10.2g/mL
    DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
    Physical Form Powder
    MDL Number MFCD00011184
    Color Dark brown
    Quantity 5g
    Concentration Pt 84.4% min.
    Chemical Name or Material Platinum(IV) oxide, Anhydrous
    Assay 99.95%, (metals basis)
    Grade Premion™

    GHS H Statement
    H272
    May intensify fire
    oxidizer.

    GHS P Statement
    P221-P210-P220-P280-P370+P378a-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Keep/Store away from clothing/combustible materials.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit