Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Platinum(IV) oxide hydrate, 99.9% (metals basis), Pt 71-75%

Manufacturer:  Alfa Aesar™ 1104901

Quantity 0.25g

 View more versions of this product

Catalog No. AA1104901


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

52785-06-5
227.09 (Anhydrous)
UN1479
Platinum(IV) oxide hydrate
PtO2·xH2O
MFCD00066964
14,7527

Specifications

52785-06-5
PtO2·xH2O
MFCD00066964
14,7527
Dark brown
Platinum(IV) oxide hydrate
Powder
227.09 (Anhydrous)
UN1479
Pt 71 to 75%
0.25g
99.9%, (metals basis)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H272
May intensify fire
oxidizer.

GHS P Statement
P221-P210-P220-P280-P370+P378a-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.

RTECSNumber : TP2506020

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit