Alfa Aesar™ Platinum, nominally 70% on high surface area advanced carbon support, HiSPEC™ 13100

Manufacturer: Alfa Aesar™ 4504106

Quantity 5g

 View more versions of this product

Catalog No. AA4504106

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00011179
    UN Number UN1325
    Chemical Name or Material Platinum, HiSPEC™ 13100, Nominally 70% on high surface area advanced carbon support

    Specifications

    Health Hazard 3 GHS P Statement
    P210-P280-P281-P308+P313-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    UN Number UN1325
    Health Hazard 2 GHS H Statement
    H228-H351
    Flammable solid.
    Suspected of causing cancer.
    Color Dark gray to black
    Quantity 5g
    DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
    MDL Number MFCD00011179
    Odor Odorless
    Health Hazard 1 Danger
    Chemical Name or Material Platinum, HiSPEC™ 13100, Nominally 70% on high surface area advanced carbon support

    GHS H Statement
    H228-H351
    Flammable solid.
    Suspected of causing cancer.

    GHS P Statement
    P210-P280-P281-P308+P313-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit