Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Platinum Plasma Std. Solution, Alfa Aesar™, Specpure™

Manufacturer:  Alfa Aesar 14397AP

Quantity 500mL

 View more versions of this product

Catalog No. AA14397AP


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

Liquid
Pt in 20% HCl
MFCD00011179
Platinum

Specifications

Liquid
Pt in 20% HCl
Characteristic
Platinum
Liquid
MFCD00011179
500mL

Safety and Handling

SDS & Safety Documents

H314-H335: Causes severe skin burns and eye damage. May cause respiratory irritation.

DOTInformation : Hazard Class: 8, Packing Group: II

EINECSNumber : 231-595-7

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit