Alfa Aesar™ Platinum powder, -200+400 mesh, 99.98% (metals basis)

Manufacturer: Alfa Aesar™ 4683006

Quantity 5g

 View more versions of this product

Catalog No. AA4683006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-06-4
    MDL Number MFCD0001117
    Chemical Name or Material Platinum powder

    Specifications

    TSCA TSCA
    Mesh Size −200+400 Mesh
    MDL Number MFCD0001117
    CAS 7440-06-4
    Quantity 5g
    RTECS Number TP2160000
    Odor Odorless
    Physical Form Powder
    Chemical Name or Material Platinum powder
    Assay 99.98%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit