Learn More
Invitrogen™ eBioscience™ Platinum ProcartaPlex Human Basic Kit for Serum Samples Non-distribution product as customer accommodation.
Bead-based multiplexing assay system for quantificaton of multiple protein biomarkers in a single sample. Newly developed matrix-type-specific diluents for plasma and serum overcome matrix effects and assure high performance specifications (e.g. spike and dilution recovery results 70-130%)
Manufacturer:  Invitrogen™ EPXP01010422901
Catalog No. 50-112-5893
Add to cart
Includes: Platinum Human Standard Mix P1
Platinum Human Control P1 High
Platinum Human Control P1 Low
Reading Buffer
Wash Buffer 10x
Streptavidin-PE
Assay Diluent for Serum Samples
Detection Antibody Diluent Human
PCR 8-Tube Strip
Adhesive Film
Greiner Microplate black 96 well
Description
Description
  • Platinum ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
  • Basic kits are designed for the use with Platinum Simplex kits
  • This kit includes antigen standards, controls, diluents, wash buffers, SA-PE a plates
  • Only one basic kit is needed to run a panel of Simplex kits
Specifications
Specifications
Multiplex Immunoassay
Serum, Plasma
Human
96 tests
Serum, Plasma: 25μL