Learn More
Invitrogen™ eBioscience™ Platinum ProcartaPlex Human IL-10 Simplex Non-distribution product as customer accommodation.
Bead-based multiplexing assay system for quantificaton of multiple protein biomarkers in a single sample. Newly developed matrix-type-specific diluents for plasma and serum overcome matrix effects and assure high performance specifications (e.g. spike and dilution recovery results 70-130%)
Manufacturer:  Invitrogen™ EPXP01010215901
Catalog No. 50-112-5872
Add to cart
Includes: Antibody Magnetic Beads (50X) coated with monoclonal antibody to human IL-10
Biotin-Conjugate (50x) anti-human IL-10 monoclonal antibody
Description
Description
  • Platinum ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
  • Platinum ProcartaPlex Simplex kits can only be combined with one another
Specifications
Specifications
Multiplex Immunoassay
Serum, Plasma
Human
96 tests
Serum, Plasma: 25μL
Interleukin-10, TGIF, MCSF III, CSIF, B TCGF