Learn More
Invitrogen™ eBioscience™ Platinum ProcartaPlex Human Panel 1 (42 plex) Non-distribution product as customer accommodation.
Bead-based multiplexing assay system for quantificaton of multiple protein biomarkers in a single sample. Newly developed matrix-type-specific diluents for plasma and serum overcome matrix effects and assure high performance specifications (e.g. spike and dilution recovery results 70-130%)
Manufacturer:  Invitrogen™ EPXP42010200901
Catalog No. 50-112-5912
Add to cart
Includes: Platinum Human Standard Mix P1
Platinum Human Control P1 High
Platinum Human Control P1 Low
Platinum Human Bead Mix
Platinum Human Detection Antibody Mix
Reading Buffer
Wash Buffer 10x
Streptavidin-PE
Assay Diluent for Plasma Samples
Assay Diluent for Serum Samples
PCR 8-Tube Strip
Adhesive Film
Greiner Microplate black 96 well
Description
Description
  • Platinum ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
  • Platinum ProcartaPlex Panels are ready-to-use kits and are not combinable with other Platinum ProcartaPlex Simplex kits or kits from different format
Specifications
Specifications
Multiplex Immunoassay
Serum, Plasma
Human
96 tests
Serum, Plasma: 25μL
Brain-Derived Neurotrophic Factor (BDNF), CCL11 (Eotaxin), CCL3 (MIP-1 alpha), CCL4 (MIP-1 beta), CCL5 (RANTES), CCL8 (MCP-2), CD31 (PECAM-1), CD62P (P-Selectin), CXCL1 (GRO alpha), CXCL10 (IP-10), GM-CSF, HGF, IFN alpha, IFN gamma, IL-1 alpha, IL-1 beta, IL-1 Receptor Antagonist, IL-10, IL-12, IL-13, IL-15, IL-16, IL-17A, IL-2, IL-20, IL-21, IL-4, IL-5, IL-6, IL-7, IL-8 (CXCL8), IL-9, LIF, OPG, PDGF-BB, sCD120b (sTNF Receptor II), Stem Cell Factor (SCF), t-PA, TNF alpha, TSLP, VEGF-A, VEGF-D