Learn More
Invitrogen™ eBioscience™ Platinum ProcartaPlex Human Panel 2 (13 plex) Non-distribution product as customer accommodation.
Bead-based multiplexing assay system for quantificaton of multiple protein biomarkers in a single sample. Newly developed matrix-type-specific diluents for plasma and serum overcome matrix effects and assure high performance specifications (e.g. spike and dilution recovery results 70-130%)
Manufacturer:  Invitrogen™ EPXP13010100901
Catalog No. 50-112-5911
Add to cart
Includes: Platinum Human Standard Mix P1
Platinum Human Control P1 High
Platinum Human Control P1 Low
Platinum Human Panel 2 Bead Mix
Platinum Human Panel 2 Detection Antibody Mix
Reading Buffer
Wash Buffer 10x
Streptavidin-PE
Assay Diluent for Plasma Samples
Assay Diluent for Serum Samples
PCR 8-Tube Strip
Adhesive Film
Greiner Microplate black 96 well
Description
Description
  • Platinum ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
  • Platinum ProcartaPlex Panels are ready-to-use kits and are not combinable with other Platinum ProcartaPlex Simplex kits or kits from different format
Specifications
Specifications
Multiplex Immunoassay
Serum, Plasma
Human
96 tests
Serum, Plasma: 25μL
GM-CSF, IFN gamma, IL-1 beta, IL-10, IL-12, IL-13, IL-17A, IL-2, IL-4, IL-5, IL-6, IL-8 (CXCL8), TNF alpha