Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Platinum Rhodium foil, 0.025mm (0.001 in.) thick, 99.9% (metals basis), Alfa Aesar™

Used as a standard for calibration of base metal thermocouples. It is also used in the glass industry for furnace linings and complex fabrications.

$175.40 - $1951.00

Specifications

MDL Number MFCD00198186
Chemical Name or Material Platinum Rhodium foil
Molecular Formula Pt:Rh; 80:20 wt%
Packaging Tube
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AA41811FF View Documents Alfa Aesar
41811FF
25 x 25mm Each for $175.40
Order in Bulk

Request bulk or custom formats

Add to cart
 
AA41811FI View Documents Alfa Aesar
41811FI
50 x 50mm Each for $584.50
Order in Bulk

Request bulk or custom formats

Add to cart
 
AA41811GH View Documents Alfa Aesar
41811GH
100 x 100mm Each for $1,951.00
Order in Bulk

Request bulk or custom formats

Add to cart
 
Specifications

Specifications

MFCD00198186
Platinum Rhodium foil
Pt:Rh; 80:20 wt%
Tube
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit